Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
$2,066.17 - $4,070.42
Specifications
Format Half-Area
Lid With Lid
Array 8 x 12
Color Black, Clear Bottom
No. of Wells 96
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
15100170
GREENER_CHOICE
View Documents Promotion Details
Corning™
4680
4/Pk
Case of 16 Each for $2,066.17
Only null left
07200841
GREENER_CHOICE
View Documents Promotion Details
Corning™
3882
20/Pk., 100/Cs.
Case of 100 Each for $4,070.42
Only null left
Description
Description
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications
    Specifications
    Half-Area
    8 x 12
    96
    Cell Culture
    With Lid
    Black, Clear Bottom
    Sterile
    Flat Bottom (F-well)