missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate

Description
Specifications
Specifications
| Format | Half-Area |
| Lid | With Lid |
| Array | 8 x 12 |
| Packaging | 4/Pk |
| Binding Type | TC-Treated |
| Color | Black, Clear Bottom |
| For Use With (Equipment) | For high content cell-based assays using imaging systems |
| Material | Cycloolefin Copolymer |
| No. of Wells | 96 |
| Sterility | Sterile |
| Show More |