missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 9000 Series Glass Particle Standards

Replace polystyrene spheres in many applications with dry glass NIST™-traceable size standards.

$1,302.09 - $1,387.50

Specifications

Additive None
Content And Storage Store at room temperature. Keep bottle tightly sealed to avoid contamination.
Color White
For Use With (Application) Narrow Size Standard Deviation Applications
Product Type Size Standard Particles
View More Specs

Products 25
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09980083
View Documents Promotion Details
Thermo Scientific™
9002
2 μm
Each for $1,302.81
Only null left
 
09980085
View Documents Promotion Details
Thermo Scientific™
DUK9008
8 μm
Each for $1,302.09
Only null left
 
09980086
View Documents Promotion Details
Thermo Scientific™
DUK9010
10.0 ±1.0 μm
Each for $1,302.09
Only null left
 
09980087
View Documents Promotion Details
Thermo Scientific™
9015
15 μm
Each for $1,384.91
Only null left
 
09980088
View Documents Promotion Details
Thermo Scientific™
9020
20 μm
Each for $1,387.50
Only null left
 
09980089
View Documents Promotion Details
Thermo Scientific™
DUK9030
30 μm
Each for $1,302.09
Only null left
 
09980090
View Documents Promotion Details
Thermo Scientific™
DUK9040
40 μm
Each for $1,384.13
Only null left
 
09980091
View Documents Promotion Details
Thermo Scientific™
DUK9050
50 μm
Each for $1,302.09
Only null left
 
09980092
View Documents Promotion Details
Thermo Scientific™
DUK9060
60 μm
Each for $1,302.09
Only null left
 
09980093
View Documents Promotion Details
Thermo Scientific™
DUK9070
70 μm
Each for $1,302.09
Only null left
 
09980096
View Documents Promotion Details
Thermo Scientific™
9100
100 μm
Each for $1,302.81
Only null left
 
09980111
View Documents Promotion Details
Thermo Scientific™
91000
1000 μm
Each for $1,302.81
Only null left
 
09980097
View Documents Promotion Details
Thermo Scientific™
9110
110 μm
Each for $1,302.81
Only null left
 
09980099
View Documents Promotion Details
Thermo Scientific™
9140
140 μm
Each for $1,302.81
Only null left
 
09980100
View Documents Promotion Details
Thermo Scientific™
9170
170 μm
Each for $1,302.81
Only null left
 
09980101
View Documents Promotion Details
Thermo Scientific™
DUK9200
200 μm
Each for $1,302.09
Only null left
 
09980112
View Documents Promotion Details
Thermo Scientific™
92000
2000 μm
Each for $1,302.81
Only null left
 
09980102
View Documents Promotion Details
Thermo Scientific™
9230
230 μm
Each for $1,302.81
Only null left
 
09980103
View Documents Promotion Details
Thermo Scientific™
9280
280 μm
Each for $1,302.81
Only null left
 
09980104
View Documents Promotion Details
Thermo Scientific™
9330
330 μm
Each for $1,384.91
Only null left
 
09980105
View Documents Promotion Details
Thermo Scientific™
DUK9400
400 μm
Each for $1,302.09
Only null left
 
09980106
View Documents Promotion Details
Thermo Scientific™
9480
480 μm
Each for $1,302.81
Only null left
 
09980107
View Documents Promotion Details
Thermo Scientific™
9550
550 μm
Each for $1,302.81
Only null left
 
09980110
View Documents Promotion Details
Thermo Scientific™
DUK9950
950 μm
Each for $1,302.09
Only null left
 
09980084
View Documents Promotion Details
Thermo Scientific™
9005
5 μm
Each for $1,302.81
Only null left
 
Description

Description

Replace polystyrene spheres in many applications with Thermo Scientific™ 9000 Series Glass Particle Standards, dry glass NIST™-traceable size standards. Glass microspheres can be used in any application that requires a NIST™-traceable size standard with a narrow size distribution where sample conditions may not be suitable for polystyrene spheres. The glass microspheres provide a better tolerance to chemicals and solvents in addition to higher mechanical and thermal stability.

The Glass Size Standards are highly uniform borosilicate or soda lime glass spheres calibrated with NIST-traceable methodology. Standards are ideal for the calibration of microscopes, light scattering instruments and other particle-measuring equipment. They are also used in laser light scattering studies and colloidal systems research.
  • Mean diameters have been calibrated with Thermo Fisher Scientific™'s NIST™-traceable microscopy methods
  • Size distribution and uniformity are measured by optical microscopy
  • Spheres are used instead of irregularly shaped particles to minimize the response of analytical systems to shape effects
  • Products are packaged as dry spheres
  • Packages are lot-numbered for convenient technical service and support after the sale
  • 9000 Series products from 2μm to 20μm are made from borosilicate glass, have a particle density of 2.5 to 2.55g/cm3 and an index of refraction of 1.56 at 589nm (25°C)
  • Products from 30μm to 2000μm are made from soda lime glass, have a particle density of 2.4 to 2.5g/cm3 and an index of refraction of 1.51 at 589nm (25°C)

Includes:

Certificate of Calibration and Traceability to NIST™, including description of the calibration method and its uncertainty, table of chemical and physical properties, and Material Safety Data Sheet with handling and disposal instructions.

Recommended for:

Spacer applications where uniform bond thickness is a necessity.

Note:

Thermo Fisher Scientific™ has its own engineering, production and metrology facilities for better quality control and response to customer needs. In addition to this line of polymer size standards, we also offer calibration standards in silica and glass materials. Ask our technical service department for information about specific applications or other product needs not listed here.

Specifications

Specifications

None
White
Size Standard Particles
Product stable for a minimum of 24 months
Store at room temperature. Keep bottle tightly sealed to avoid contamination.
Narrow Size Standard Deviation Applications
1 g
Videos
SDS
Documents

Documents