missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole 98.0+%, TCI America™

Supplier:  TCI America P149225G

 View more versions of this product

Catalog No. P149225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

9-Phenylcarbazole
88°C
C18H13N
25 g
VIJYEGDOKCKUOL-UHFFFAOYSA-N
9-phenyl-9H-carbazole
70851
≥98.0% (GC)
1150-62-5
White-Yellow
MFCD00004965
n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
243.31
243.31
Crystalline Powder

Chemical Identifiers

1150-62-5
243.31
VIJYEGDOKCKUOL-UHFFFAOYSA-N
70851
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
C18H13N
MFCD00004965
n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
9-phenyl-9H-carbazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions