missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole 98.0+%, TCI America™

$36.57 - $437.02

Chemical Identifiers

CAS 1150-62-5
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00004965
InChI Key VIJYEGDOKCKUOL-UHFFFAOYSA-N
Synonym n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
PubChem CID 70851
IUPAC Name 9-phenyl-9H-carbazole
SMILES C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P14921G
View Documents
TCI America
P14921G
1 g N/A
Only null left
 
P14925G
View Documents
TCI America
P14925G
5 g N/A
Only null left
 
P149225G
View Documents
TCI America
P149225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1150-62-5
243.31
VIJYEGDOKCKUOL-UHFFFAOYSA-N
70851
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
C18H13N
MFCD00004965
n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
9-phenyl-9H-carbazole

Specifications

1150-62-5
White-Yellow
MFCD00004965
n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
243.31
243.31
Crystalline Powder
88°C
C18H13N
1 g
VIJYEGDOKCKUOL-UHFFFAOYSA-N
9-phenyl-9H-carbazole
70851
≥98.0% (GC)
9-Phenylcarbazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'