missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylanthracene 98.0+%, TCI America™

Catalog No. p-7134443
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01381G 1 g
P01385G 5 g
P013825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P01381G Supplier TCI America Supplier No. P01381G
Only null left

Chemical Identifiers

CAS 602-55-1
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00001252
InChI Key LUBXLGUQZVKOFP-UHFFFAOYSA-N
Synonym anthracene, 9-phenyl, 9-phenyl-anthracene, 10-phenylanthracene, 9-phenyl-anthracen, 9-phenylanthracene, acmc-1b14f, anthracene, 9-phenyl-8ci 9ci
PubChem CID 11766
IUPAC Name 9-phenylanthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 154°C
Quantity 1 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenylanthracene