missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-9-fluorenol 98.0+%, TCI America™

Catalog No. P12315G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12315G Supplier TCI America Supplier No. P12315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25603-67-2
Molecular Formula C19H14O
Molecular Weight (g/mol) 258.32
MDL Number MFCD00037121
InChI Key UJPHBDAPVWFPTG-UHFFFAOYSA-N
Synonym 9-Hydroxy-9-phenylfluorene
PubChem CID 141227
IUPAC Name 9-phenylfluoren-9-ol
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)O

Specifications

Melting Point 111°C
Color Yellow
Quantity 5 g
Formula Weight 258.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-9-fluorenol