missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™

Supplier:  TCI America P23501G

 View more versions of this product

Catalog No. P23501G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
240°C
C30H35B2NO4
9-Phenyl-9H-carbazole-3,6-diboronic Acid Bis(pinacol) Ester
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=C3C=C(C=C4)B5OC(C(O5)(C)C)(C)C)C6=CC=CC=C6
495.233
495.23
Crystalline Powder
618442-57-2
White-Yellow
1 g
BMKVLWGCSCKZTD-UHFFFAOYSA-N
9-phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
12177223
≥98.0% (HPLC,N)

Chemical Identifiers

618442-57-2
495.233
9-Phenyl-9H-carbazole-3,6-diboronic Acid Bis(pinacol) Ester
9-phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
C30H35B2NO4
BMKVLWGCSCKZTD-UHFFFAOYSA-N
12177223
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=C3C=C(C=C4)B5OC(C(O5)(C)C)(C)C)C6=CC=CC=C6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions