missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™

Catalog No. p-7135535
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2350200MG 200 mg
P23501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2350200MG Supplier TCI America Supplier No. P2350200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 618442-57-2
Molecular Formula C30H35B2NO4
Molecular Weight (g/mol) 495.233
InChI Key BMKVLWGCSCKZTD-UHFFFAOYSA-N
Synonym 9-Phenyl-9H-carbazole-3,6-diboronic Acid Bis(pinacol) Ester
PubChem CID 12177223
IUPAC Name 9-phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=C3C=C(C=C4)B5OC(C(O5)(C)C)(C)C)C6=CC=CC=C6

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 200 mg
Formula Weight 495.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole