missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 209°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 779.95 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole |
Safety and Handling
| TSCA | No |