accessibility menu, dialog, popup

UserName

NNNNN

CAS RN 1258780-50-5

IUPAC Name: 9-phenyl-3,6-bis[4-(1-phenyl-1H-1,3-benzodiazol-2-yl)phenyl]-9H-carbazole
Synonyms: 9-phenyl-3,6-bis[4-(1-phenyl-1,3-benzodiazol-2-yl)phenyl]carbazole
Molecular Weight (g/mol): 779.95
Molecular Formula: C56H37N5
InChi Key: CWUOZRFIMAVZLO-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N1C2=C(C=C(C=C2)C2=CC=C(C=C2)C2=NC3=CC=CC=C3N2C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=C(C=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results