missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole 98.0+%, TCI America™

Catalog No. p-7135519
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2330200MG 200 mg
P23301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2330200MG Supplier TCI America Supplier No. P2330200MG
Only null left

Chemical Identifiers

CAS 1258780-50-5
Molecular Formula C56H37N5
Molecular Weight (g/mol) 779.947
InChI Key CWUOZRFIMAVZLO-UHFFFAOYSA-N
PubChem CID 101525816
IUPAC Name 9-phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)C5=NC6=CC=CC=C6N5C7=CC=CC=C7)C8=C2C=CC(=C8)C9=CC=C(C=C9)C1=NC2=CC=CC=C2N1C1=CC=CC=C1

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 200 mg
Formula Weight 779.95
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole