missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™

Catalog No. P23761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23761G 1 g
P23765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23761G Supplier TCI America Supplier No. P23761G

Chemical Identifiers

CAS 1126522-69-7
Molecular Formula C24H24BNO2
Molecular Weight (g/mol) 369.27
MDL Number MFCD16621140
InChI Key UBASCOPZFCGGAV-UHFFFAOYSA-N
Synonym 9-Phenylcarbazole-3-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-3-yl)-1,3,2-dioxaborolane
PubChem CID 57415695
IUPAC Name 9-phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C
Color White
Quantity 1 g
Formula Weight 369.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole