missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7135599
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25611G 1 g
P25615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25611G Supplier TCI America Supplier No. P25611G

Chemical Identifiers

CAS 1219956-30-5
Molecular Formula C30H28BNO2
Molecular Weight (g/mol) 445.369
MDL Number MFCD30187279
InChI Key AIYLNDJDCBJMMY-UHFFFAOYSA-N
Synonym 4-(9-Phenyl-9H-carbazol-3-yl)phenylboronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-[4-(9-phenylcarbazole-3-yl)phenyl]-1,3,2-dioxaborolane
PubChem CID 58340519
IUPAC Name 9-phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC4=C(C=C3)N(C5=CC=CC=C54)C6=CC=CC=C6

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 1 g
Formula Weight 445.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole