CAS RN 1219956-30-5
IUPAC Name:
9-phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole
Synonyms:
9-phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
Molecular Weight (g/mol):
445.37
Molecular Formula:
C30H28BNO2
InChi Key:
AIYLNDJDCBJMMY-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
9-Phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole 98.0+%, TCI America™
CAS: 1219956-30-5 Molecular Formula: C30H28BNO2 Molecular Weight (g/mol): 445.369 MDL Number: MFCD30187279 InChI Key: AIYLNDJDCBJMMY-UHFFFAOYSA-N Synonym: 4-(9-Phenyl-9H-carbazol-3-yl)phenylboronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-[4-(9-phenylcarbazole-3-yl)phenyl]-1,3,2-dioxaborolane PubChem CID: 58340519 IUPAC Name: 9-phenyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=CC4=C(C=C3)N(C5=CC=CC=C54)C6=CC=CC=C6