missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 96.0+%, TCI America™
Supplier: TCI America P25711G
Specifications
| 9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole | |
| 238°C | |
| C30H35B2NO4 | |
| BLKLIQJTGNDTOL-UHFFFAOYSA-N | |
| 9-phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole | |
| 90297524 | |
| ≥96.0% (HPLC,N) |
| 1035631-57-2 | |
| White-Yellow | |
| 1 g | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C5=CC=CC=C5)C=C(C=C4)B6OC(C(O6)(C)C)(C)C | |
| 495.233 | |
| 495.23 | |
| Crystalline Powder |
Chemical Identifiers
| 1035631-57-2 | |
| 495.233 | |
| 90297524 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C5=CC=CC=C5)C=C(C=C4)B6OC(C(O6)(C)C)(C)C |
| C30H35B2NO4 | |
| BLKLIQJTGNDTOL-UHFFFAOYSA-N | |
| 9-phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
Safety and Handling
TSCA : No