missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 96.0+%, TCI America™

Catalog No. p-7135603
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2571200MG 200 mg
P25711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2571200MG Supplier TCI America Supplier No. P2571200MG
Only null left

Chemical Identifiers

CAS 1035631-57-2
Molecular Formula C30H35B2NO4
Molecular Weight (g/mol) 495.233
InChI Key BLKLIQJTGNDTOL-UHFFFAOYSA-N
PubChem CID 90297524
IUPAC Name 9-phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C5=CC=CC=C5)C=C(C=C4)B6OC(C(O6)(C)C)(C)C

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 200 mg
Formula Weight 495.23
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole