Learn More
9-Phenyl-2,3,7-trihydroxy-6-fluorone
CAS: 975-17-7 | C19H12O5 | 320.3 g/mol
$102.74 - $1271.01
Chemical Identifiers
| CAS | 975-17-7 |
|---|---|
| Molecular Formula | C19H12O5 |
| Molecular Weight (g/mol) | 320.3 |
| MDL Number | MFCD00005048 |
| InChI Key | YDCFOUBAMGLLKA-UHFFFAOYSA-N |
| Synonym | phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone |
| PubChem CID | 70420 |
| IUPAC Name | 2,6,7-trihydroxy-9-phenylxanthen-3-one |
| SMILES | C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1149506
|
Thermo Scientific Chemicals
A1149506 |
5 g |
Each for $102.74
|
|
|||||
|
AAA1149514
|
Thermo Scientific Chemicals
A1149514 |
25 g |
Each for $413.08
|
|
|||||
|
AAA1149522
|
Thermo Scientific Chemicals
A1149522 |
100 g |
Each for $1,271.01
|
|
|||||
Description
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
Solubility
Soluble in chloroform.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
Chemical Identifiers
| 975-17-7 | |
| 320.3 | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 70420 | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
| C19H12O5 | |
| MFCD00005048 | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one |
Specifications
| 975-17-7 | |
| C19H12O5 | |
| 5 g | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one | |
| 70420 | |
| 9-Phenyl-2,3,7-trihydroxy-6-fluorone |
| >300°C | |
| MFCD00005048 | |
| 40910 | |
| Soluble in chloroform. | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O | |
| 320.3 | |
| 320.3 |
Safety and Handling
EINECSNumber : 213-550-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only