Learn More
9-Phenyl-2,3,7-trihydroxy-6-fluorone
CAS: 975-17-7 | C19H12O5 | 320.3 g/mol
Supplier: Thermo Scientific Chemicals A1149522
Description
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
Solubility
Soluble in chloroform.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
Specifications
| 9-Phenyl-2,3,7-trihydroxy-6-fluorone | |
| >300°C | |
| MFCD00005048 | |
| 40910 | |
| Soluble in chloroform. | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O | |
| 320.3 | |
| 320.3 |
| 975-17-7 | |
| C19H12O5 | |
| 100 g | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one | |
| 70420 |
Chemical Identifiers
| 975-17-7 | |
| 320.3 | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 70420 | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
| C19H12O5 | |
| MFCD00005048 | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one |
Safety and Handling
EINECSNumber : 213-550-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only