missing translation for 'onlineSavingsMsg'
Learn More

9-Oxoacridine-10-acetic Acid 98.0+%, TCI America™

Catalog No. p-7134336
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04321G 1 g
O04325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04321G Supplier TCI America Supplier No. O04321G

Chemical Identifiers

CAS 38609-97-1
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00157050
InChI Key UOMKBIIXHQIERR-UHFFFAOYSA-N
Synonym 10-(Carboxymethyl)-9(10H)-acridone, 9-Oxo-10(9H)-acridineacetic Acid, Cridanimod
PubChem CID 38072
IUPAC Name 2-(9-oxoacridin-10-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2CC(=O)O

Specifications

Melting Point 291°C
Color Yellow
Quantity 1 g
Formula Weight 253.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Oxoacridine-10-acetic Acid
RTECSNumber AR7190000
TSCA No