missing translation for 'onlineSavingsMsg'
Learn More

9-Octadecynoic acid, 98%

Catalog No. AAL0129406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0129406 5 g
AAL0129403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0129406 Supplier Thermo Scientific Chemicals Supplier No. L0129406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 506-24-1 | C18H32O2 | 280.45 g/mol

9-Octadecynoic acid, is used as an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Octadecynoic acid, is used as an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 506-24-1
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.45
MDL Number MFCD00014386
InChI Key RGTIBVZDHOMOKC-UHFFFAOYSA-N
Synonym stearolic acid, 9-octadecynoic acid, 9-stearolic acid, delta 9-octadecynoic acid, stearolsaeure, octadec-9-insaeure, octadeca-9-ynoic acid, 9-odya, d09qia, 9-oda
PubChem CID 68167
ChEBI CHEBI:28801
IUPAC Name octadec-9-ynoic acid
SMILES CCCCCCCCC#CCCCCCCCC(O)=O

Specifications

Melting Point 43°C to 47°C
Boiling Point 189°C to 190°C (2 mmHg)
Quantity 5 g
Beilstein 1786634
Solubility Information Insoluble in water.
Formula Weight 280.45
Percent Purity 98%
Chemical Name or Material 9-Octadecynoic acid
EINECSNumber 208-030-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only