accessibility menu, dialog, popup

UserName

CAS RN 506-24-1

H3COHO

CAS RN 506-24-1

IUPAC Name: octadec-9-ynoic acid
Synonyms: stearolic acid delta(9)-octadecynoic acid 9-stearolic acid 9-oda stearolsaeure 9-odya octadec-9-insaeure
Molecular Weight (g/mol): 280.45
Molecular Formula: C18H32O2
InChi Key: RGTIBVZDHOMOKC-UHFFFAOYSA-N
SMILES: CCCCCCCCC#CCCCCCCCC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results