missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Methylcarbazole 99.0+%, TCI America™
$130.48 - $385.86
Chemical Identifiers
| CAS | 1484-12-4 |
|---|---|
| Molecular Formula | C13H11N |
| Molecular Weight (g/mol) | 181.238 |
| MDL Number | MFCD00013431 |
| InChI Key | SDFLTYHTFPTIGX-UHFFFAOYSA-N |
| Synonym | n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole |
| PubChem CID | 15142 |
| IUPAC Name | 9-methylcarbazole |
| SMILES | CN1C2=CC=CC=C2C3=CC=CC=C31 |
Chemical Identifiers
| 1484-12-4 | |
| 181.238 | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 15142 | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 |
| C13H11N | |
| MFCD00013431 | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| 9-methylcarbazole |
Specifications
| 1484-12-4 | |
| 195°C | |
| MFCD00013431 | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 | |
| 181.238 | |
| 181.24 | |
| Crystalline Powder |
| 89°C | |
| C13H11N | |
| 5 g | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 9-methylcarbazole | |
| 15142 | |
| ≥99.0% (GC) | |
| 9-Methylcarbazole |
Safety and Handling
RTECSNumber : FE6266000
TSCA : No