missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Methylcarbazole 99.0+%, TCI America™
Supplier: TCI America M096025G
Specifications
| 9-Methylcarbazole | |
| 89°C | |
| C13H11N | |
| 25 g | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 9-methylcarbazole | |
| 15142 | |
| ≥99.0% (GC) |
| 1484-12-4 | |
| 195°C | |
| MFCD00013431 | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 | |
| 181.238 | |
| 181.24 | |
| Crystalline Powder |
Chemical Identifiers
| 1484-12-4 | |
| 181.238 | |
| SDFLTYHTFPTIGX-UHFFFAOYSA-N | |
| 15142 | |
| CN1C2=CC=CC=C2C3=CC=CC=C31 |
| C13H11N | |
| MFCD00013431 | |
| n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole | |
| 9-methylcarbazole |
Safety and Handling
RTECSNumber : FE6266000
TSCA : No