missing translation for 'onlineSavingsMsg'
Learn More

9-Mesityl-10-methylacridinium Perchlorate 98.0+%, TCI America™

Catalog No. m17745g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17745G 5 g
M17741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17745G Supplier TCI America Supplier No. M17745G
Only null left

Chemical Identifiers

CAS 674783-97-2
Molecular Formula C23H22ClNO4
Molecular Weight (g/mol) 411.882
InChI Key LFMBERYWDLWXNO-UHFFFAOYSA-M
Synonym 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium Perchlorate
PubChem CID 15953479
IUPAC Name 10-methyl-9-(2,4,6-trimethylphenyl)acridin-10-ium;perchlorate
SMILES CC1=CC(=C(C(=C1)C)C2=C3C=CC=CC3=[N+](C4=CC=CC=C42)C)C.[O-]Cl(=O)(=O)=O

Specifications

Color Yellow
Quantity 5 g
UN Number 1479
Formula Weight 411.88
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Mesityl-10-methylacridinium Perchlorate