missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 9-Hydroxyrisperidone, 98%

Catalog No. AC456250010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456250010 Supplier Thermo Scientific Chemicals Supplier No. 456250010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144598-75-4 | C23H27FN4O3 | 426.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144598-75-4
Molecular Formula C23H27FN4O3
Molecular Weight (g/mol) 426.48
InChI Key PMXMIIMHBWHSKN-UHFFFAOYSA-N
Synonym paliperidone, 9-hydroxyrisperidone, invega, 9-oh-risperidone, invega sustenna, xeplion, palliperidone, 3-2-4-6-fluoro-1,2-benzisoxazol-3-yl-1-piperidinyl ethyl-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4h-pyrido 1,2-a pyrimidin-4-one, chembl1621, 3-2-4-6-fluorobenzo d isoxazol-3-yl piperidin-1-yl ethyl-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4h-pyrido 1,2-a pyrimidin-4-one
PubChem CID 115237
ChEBI CHEBI:83804
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCC(C2=N1)O)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Melting Point 171.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material 9-Hydroxyrisperidone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only