missing translation for 'onlineSavingsMsg'
Learn More

9-Hydroxyfluorene-9-carboxylic Acid 98.0+%, TCI America™

Catalog No. H100425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H100425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H100425G Supplier TCI America Supplier No. H100425G
Only null left

Chemical Identifiers

CAS 467-69-6
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00001137
InChI Key GXAMYUGOODKVRM-UHFFFAOYSA-N
Synonym flurenol, 9-hydroxy-9-fluorenecarboxylic acid, flurecol, 9-hydroxy-9h-fluorene-9-carboxylic acid, glycolic acid, diphenylene, 9h-fluorene-9-carboxylic acid, 9-hydroxy, flurecol bsi:iso, unii-4tkt65am8p, 9-hydooxyfluorene-9-carboxylic acid, 9-hydroxy-9-fluorene carboxyli acid
PubChem CID 10087
ChEBI CHEBI:82015
IUPAC Name 9-hydroxyfluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C(=O)O)O

Specifications

Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 226.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Hydroxyfluorene-9-carboxylic Acid
RTECSNumber LL6480000
TSCA Yes