missing translation for 'onlineSavingsMsg'
Learn More

9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7131139
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16421G 1 g
H16425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16421G Supplier TCI America Supplier No. H16421G

Chemical Identifiers

CAS 871696-12-7
Molecular Formula C30H43B2NO4
Molecular Weight (g/mol) 503.297
MDL Number MFCD30187269
InChI Key VUJPSYQJTJSPOI-UHFFFAOYSA-N
PubChem CID 25150161
IUPAC Name 9-hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3CCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 223°C
Color White
Quantity 1 g
Formula Weight 503.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole