missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenylmethyl Carbazate 98.0+%, TCI America™
$102.52 - $268.43
Chemical Identifiers
| CAS | 35661-51-9 |
|---|---|
| Molecular Formula | C15H14N2O2 |
| Molecular Weight (g/mol) | 254.29 |
| MDL Number | MFCD00239419 |
| InChI Key | YGCGPEUVGHDMLO-UHFFFAOYSA-N |
| Synonym | Carbazic Acid 9-Fluorenylmethyl Ester, Fmoc-hydrazine, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine |
| PubChem CID | 161883 |
| IUPAC Name | [(9H-fluoren-9-yl)methoxy]carbohydrazide |
| SMILES | NNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Chemical Identifiers
| 35661-51-9 | |
| 254.29 | |
| YGCGPEUVGHDMLO-UHFFFAOYSA-N | |
| 161883 | |
| NNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C15H14N2O2 | |
| MFCD00239419 | |
| Carbazic Acid 9-Fluorenylmethyl Ester, Fmoc-hydrazine, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine | |
| [(9H-fluoren-9-yl)methoxy]carbohydrazide |
Specifications
| 35661-51-9 | |
| White | |
| MFCD00239419 | |
| Carbazic Acid 9-Fluorenylmethyl Ester, Fmoc-hydrazine, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine | |
| NNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 254.29 | |
| 254.29 | |
| Crystalline Powder |
| 171°C | |
| C15H14N2O2 | |
| 5 g | |
| YGCGPEUVGHDMLO-UHFFFAOYSA-N | |
| [(9H-fluoren-9-yl)methoxy]carbohydrazide | |
| 161883 | |
| ≥98.0% (HPLC,T) | |
| 9-Fluorenylmethyl Carbazate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator