missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethanol 98.0+%, TCI America™

Catalog No. p-7129533
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F02005G 5 g
F020025G 25 g
2 options

Catalog No. F02005G

Supplier: TCI America F02005G

Only null left

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00001139
InChI Key XXSCONYSQQLHTH-UHFFFAOYSA-N
Synonym 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
PubChem CID 90466
IUPAC Name (9H-fluoren-9-yl)methanol
SMILES OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethanol