Learn More
9-Fluorenylmethanol, 99%
CAS: 24324-17-2 | C14H12O | 196.25 g/mol
Supplier: Thermo Scientific Chemicals A1521214
Description
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 9-Fluorenylmethanol | |
| 102°C to 106°C | |
| MFCD00001139 | |
| 2330017 | |
| Soluble in methanol. | |
| OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 196.25 | |
| 196.25 |
| 24324-17-2 | |
| C14H12O | |
| 25 g | |
| 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol | |
| XXSCONYSQQLHTH-UHFFFAOYSA-N | |
| 9H-fluoren-9-ylmethanol | |
| 90466 | |
| 99% |
Chemical Identifiers
| 24324-17-2 | |
| 196.25 | |
| XXSCONYSQQLHTH-UHFFFAOYSA-N | |
| 90466 |
| C14H12O | |
| MFCD00001139 | |
| 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol | |
| OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-167-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only