missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethanol, 99%

Catalog No. AAA1521222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1521222 100 g
AAA1521206 5 g
AAA1521214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1521222 Supplier Thermo Scientific Chemicals Supplier No. A1521222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24324-17-2 | C14H12O | 196.25 g/mol

9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00001139
InChI Key XXSCONYSQQLHTH-UHFFFAOYSA-N
Synonym 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
PubChem CID 90466
SMILES OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 102°C to 106°C
Quantity 100 g
Beilstein 2330017
Solubility Information Soluble in methanol.
IUPAC Name 9H-fluoren-9-ylmethanol
Formula Weight 196.25
Percent Purity 99%
Chemical Name or Material 9-Fluorenylmethanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-167-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only