missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenone 98.0+%, TCI America™
Supplier: TCI America F0021100G
Specifications
| 9-Fluorenone | |
| 84°C | |
| 342°C | |
| MFCD00001141 | |
| 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=O | |
| 180.206 | |
| CHEBI:17922 | |
| ≥98.0% (GC) |
| 486-25-9 | |
| Yellow | |
| C13H8O | |
| 100 g | |
| YLQWCDOCJODRMT-UHFFFAOYSA-N | |
| fluoren-9-one | |
| 10241 | |
| 180.21 | |
| Crystalline Powder |
Chemical Identifiers
| 486-25-9 | |
| 180.206 | |
| YLQWCDOCJODRMT-UHFFFAOYSA-N | |
| 10241 | |
| fluoren-9-one |
| C13H8O | |
| MFCD00001141 | |
| 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
| CHEBI:17922 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=O |
Safety and Handling
EINECSNumber : (4)-1618
RTECSNumber : LL8925000
TSCA : Yes