missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone 98.0+%, TCI America™

Catalog No. F0021500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0021500G 500 g
F002125G 25 g
F0021100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0021500G Supplier TCI America Supplier No. F0021500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 486-25-9
Molecular Formula C13H8O
Molecular Weight (g/mol) 180.206
MDL Number MFCD00001141
InChI Key YLQWCDOCJODRMT-UHFFFAOYSA-N
Synonym 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
PubChem CID 10241
ChEBI CHEBI:17922
IUPAC Name fluoren-9-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

Specifications

Melting Point 84°C
Color Yellow
Boiling Point 342°C
Quantity 500 g
Formula Weight 180.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone
EINECSNumber (4)-1618
RTECSNumber LL8925000
TSCA Yes