missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenone Oxime 97.0+%, TCI America™
$32.72 - $46.20
Chemical Identifiers
| CAS | 2157-52-0 |
|---|---|
| Molecular Formula | C13H9NO |
| Molecular Weight (g/mol) | 195.221 |
| MDL Number | MFCD00016356 |
| InChI Key | CRNNFEKVPRFZKJ-UHFFFAOYSA-N |
| Synonym | 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim |
| PubChem CID | 16543 |
| IUPAC Name | N-fluoren-9-ylidenehydroxylamine |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO |
Chemical Identifiers
| 2157-52-0 | |
| 195.221 | |
| CRNNFEKVPRFZKJ-UHFFFAOYSA-N | |
| 16543 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO |
| C13H9NO | |
| MFCD00016356 | |
| 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim | |
| N-fluoren-9-ylidenehydroxylamine |
Specifications
| 2157-52-0 | |
| Yellow | |
| MFCD00016356 | |
| 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO | |
| 195.221 | |
| 195.22 | |
| Crystalline Powder |
| 194°C | |
| C13H9NO | |
| 1 g | |
| CRNNFEKVPRFZKJ-UHFFFAOYSA-N | |
| N-fluoren-9-ylidenehydroxylamine | |
| 16543 | |
| ≥97.0% (GC,N) | |
| 9-Fluorenone Oxime |
Safety and Handling
RTECSNumber : LL9093000
TSCA : No