missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone Oxime 97.0+%, TCI America™

Catalog No. F00235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00235G 5 g
F00231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00235G Supplier TCI America Supplier No. F00235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2157-52-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00016356
InChI Key CRNNFEKVPRFZKJ-UHFFFAOYSA-N
Synonym 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim
PubChem CID 16543
IUPAC Name N-fluoren-9-ylidenehydroxylamine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO

Specifications

Melting Point 194°C
Color Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone Oxime
RTECSNumber LL9093000
TSCA No