missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenone oxime, 98+%, Thermo Scientific Chemicals
$69.56 - $69.56
Chemical Identifiers
| CAS | 2157-52-0 |
|---|---|
| Molecular Formula | C13H9NO |
| Molecular Weight (g/mol) | 195.221 |
| MDL Number | MFCD00016356 |
| InChI Key | CRNNFEKVPRFZKJ-UHFFFAOYSA-N |
| Synonym | 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim |
| PubChem CID | 16543 |
| IUPAC Name | N-fluoren-9-ylidenehydroxylamine |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0395006
|
Thermo Scientific Chemicals
L0395006 |
5 g |
Each for $69.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2157-52-0 | |
| 195.221 | |
| CRNNFEKVPRFZKJ-UHFFFAOYSA-N | |
| 16543 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO |
| C13H9NO | |
| MFCD00016356 | |
| 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim | |
| N-fluoren-9-ylidenehydroxylamine |
Specifications
| 2157-52-0 | |
| C13H9NO | |
| 5 g | |
| 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO | |
| 195.221 | |
| 195.22 | |
| 9-Fluorenone oxime |
| 193°C to 194°C | |
| MFCD00016356 | |
| 1871046 | |
| CRNNFEKVPRFZKJ-UHFFFAOYSA-N | |
| N-fluoren-9-ylidenehydroxylamine | |
| 16543 | |
| ≥98% |
Safety and Handling
EINECSNumber : 218-471-8
RTECSNumber : LL9093000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only