missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone oxime, 98+%, Thermo Scientific Chemicals

Catalog No. AAL0395006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0395006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0395006 Supplier Thermo Scientific Chemicals Supplier No. L0395006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2157-52-0 | Molecular Formula: C13H9NO | Molecular Weight: 195.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2157-52-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00016356
InChI Key CRNNFEKVPRFZKJ-UHFFFAOYSA-N
Synonym 9-fluorenone oxime, 9h-fluoren-9-one oxime, fluorenone oxime, 9-oximinofluorene, fluorenone-9-oxime, 9h-fluoren-9-one, oxime, fluoren-9-one, oxime, fluoren-9-one oxime, 9-hydroxyimino fluorene, fluorenonoxim
PubChem CID 16543
IUPAC Name N-fluoren-9-ylidenehydroxylamine
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=NO

Specifications

Melting Point 193°C to 194°C
Quantity 5 g
Beilstein 1871046
Formula Weight 195.22
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone oxime
EINECSNumber 218-471-8
RTECSNumber LL9093000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only