Learn More
9-Fluorenone, 98+%
CAS: 486-25-9 | C13H8O | 180.206 g/mol
Supplier: Thermo Scientific Chemicals A129100E
Description
9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.
Solubility
Soluble in water, alcohol, acetone, ether and benzene.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 9-Fluorenone | |
| 82°C to 85°C | |
| 342°C | |
| 98% | |
| MFCD00001141 | |
| 1636531 | |
| Soluble in water,alcohol,acetone,ether and benzene. | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=O | |
| 180.206 | |
| CHEBI:17922 | |
| ≥98% |
| 486-25-9 | |
| 0.9 | |
| 163°C (325°F) | |
| C13H8O | |
| 2500 g | |
| 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
| YLQWCDOCJODRMT-UHFFFAOYSA-N | |
| fluoren-9-one | |
| 10241 | |
| 180.21 |
Chemical Identifiers
| 486-25-9 | |
| 180.206 | |
| YLQWCDOCJODRMT-UHFFFAOYSA-N | |
| 10241 | |
| fluoren-9-one |
| C13H8O | |
| MFCD00001141 | |
| 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
| CHEBI:17922 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2=O |
Safety and Handling
EINECSNumber : 207-630-7
RTECSNumber : LL8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only