missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. F05901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05901G 1 g
F05905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05901G Supplier TCI America Supplier No. F05901G

Chemical Identifiers

CAS 1573-92-8
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.22
MDL Number MFCD00011537
InChI Key CBEFMGJHEKAMNI-UHFFFAOYSA-N
Synonym 9-oxo-9h-fluorene-1-carboxylic acid, 9-fluorenone-1-carboxylic acid, 9-oxo-9h-fluorenecarboxylic acid, 9-oxofluorenecarboxylic acid, 9-fluorenone-1-carboxylicacid, 9h-fluorene-1-carboxylic acid, 9-oxo, maybridge1_002192, 9-oxo-1-fluorenecarboxylic acid, 9-oxo-9h-fluorene-1-carboxylicacid, 9h-fluorenecarboxylic acid, 9-oxo
PubChem CID 74083
IUPAC Name 9-oxo-9H-fluorene-1-carboxylic acid
SMILES OC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

Specifications

Melting Point 198°C
Quantity 1 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone-1-carboxylic Acid