missing translation for 'onlineSavingsMsg'
Learn More

9-Ethynyl-9-fluorenol, 98%

Catalog No. AAL0971706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0971706 5 g
AAL0971703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0971706 Supplier Thermo Scientific Chemicals Supplier No. L0971706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13461-74-0 | C15H10O | 206.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13461-74-0
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.244
MDL Number MFCD00041555
InChI Key MMZVVJGCZZAWBN-UHFFFAOYSA-N
Synonym 9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
PubChem CID 518771
IUPAC Name 9-ethynylfluoren-9-ol
SMILES C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O

Specifications

Melting Point 109°C to 110°C
Quantity 5 g
Beilstein 2104685
Formula Weight 206.25
Percent Purity 98%
Chemical Name or Material 9-Ethynyl-9-fluorenol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only