missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole-3-carboxaldehyde N-Butyl-N-phenylhydrazone 98.0+%, TCI America™

Catalog No. E05751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05751G Supplier TCI America Supplier No. E05751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88107-84-0
Molecular Formula C25H27N3
Molecular Weight (g/mol) 369.51
MDL Number MFCD06797101
InChI Key WUJYNCPGAXBZMF-UHFFFAOYSA-N
Synonym 9-Ethyl-3-(N-butyl-N-phenylhydrazonomethyl)carbazole
PubChem CID 93978906
IUPAC Name 3-[(2-butyl-2-phenylhydrazin-1-ylidene)methyl]-9-ethyl-9H-carbazole
SMILES CCCCN(N=CC1=CC2=C(C=C1)N(CC)C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 369.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Ethylcarbazole-3-carboxaldehyde N-Butyl-N-phenylhydrazone