accessibility menu, dialog, popup

UserName

CH3NNNH3C(E/Z)

CAS RN 88107-84-0

IUPAC Name: 3-[(2-butyl-2-phenylhydrazin-1-ylidene)methyl]-9-ethyl-9H-carbazole
Synonyms: 3-[(2-butyl-2-phenylhydrazin-1-ylidene)methyl]-9-ethylcarbazole
Molecular Weight (g/mol): 369.51
Molecular Formula: C25H27N3
InChi Key: WUJYNCPGAXBZMF-UHFFFAOYSA-N
SMILES: CCCCN(N=CC1=CC2=C(C=C1)N(CC)C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results