missing translation for 'onlineSavingsMsg'
Learn More

9-Ethylcarbazole-3-carboxaldehyde N-Benzyl-N-phenylhydrazone 98.0+%, TCI America™

Catalog No. e05741g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05741G Supplier TCI America Supplier No. E05741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75238-79-8
Molecular Formula C28H25N3
Molecular Weight (g/mol) 403.529
MDL Number MFCD05453514
InChI Key RPHJRJPXKZMFFQ-BRPDVVIDSA-N
Synonym 9-Ethyl-3-(N-benzyl-N-phenylhydrazonomethyl)carbazole
PubChem CID 67330729
IUPAC Name N-benzyl-N-[(Z)-(9-ethylcarbazol-3-yl)methylideneamino]aniline
SMILES CCN1C2=C(C=C(C=C2)C=NN(CC3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C51

Specifications

Color Yellow
Quantity 1 g
Formula Weight 403.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Ethylcarbazole-3-carboxaldehyde N-Benzyl-N-phenylhydrazone