accessibility menu, dialog, popup

UserName

CAS RN 75238-79-8

IUPAC Name: 3-[(2-benzyl-2-phenylhydrazin-1-ylidene)methyl]-9-ethyl-9H-carbazole
Synonyms: 3-[(2-benzyl-2-phenylhydrazin-1-ylidene)methyl]-9-ethylcarbazole
Molecular Weight (g/mol): 403.53
Molecular Formula: C28H25N3
InChi Key: RPHJRJPXKZMFFQ-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(C=NN(CC1=CC=CC=C1)C1=CC=CC=C1)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results