missing translation for 'onlineSavingsMsg'
Learn More

9-Decyn-1-ol 94.0+%, TCI America™

Catalog No. p-7127878
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37105G 5 g
D371025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37105G Supplier TCI America Supplier No. D37105G
Only null left

Chemical Identifiers

CAS 17643-36-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00049199
InChI Key KOAOUMQUUZNRLL-UHFFFAOYSA-N
Synonym 10-Hydroxy-1-decyne
PubChem CID 543752
IUPAC Name dec-9-yn-1-ol
SMILES C#CCCCCCCCCO

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 154.25
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-Decyn-1-ol