accessibility menu, dialog, popup

UserName

HOCH

CAS RN 17643-36-6

IUPAC Name: dec-9-yn-1-ol
Synonyms: dec-9-yn-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: KOAOUMQUUZNRLL-UHFFFAOYSA-N
SMILES: OCCCCCCCCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results