missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Decen-1-ol 97.0+%, TCI America™
$108.21 - $797.43
Chemical Identifiers
| CAS | 13019-22-2 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00002992 |
| InChI Key | QGFSQVPRCWJZQK-UHFFFAOYSA-N |
| Synonym | 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol |
| PubChem CID | 25612 |
| IUPAC Name | dec-9-en-1-ol |
| SMILES | C=CCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D189225ML
|
TCI America
D189225ML |
25 mL |
Each for $108.21
|
|
|||||
|
D1892500ML
|
TCI America
D1892500ML |
500 mL |
Each for $797.43
|
|
|||||
Chemical Identifiers
| 13019-22-2 | |
| 156.269 | |
| QGFSQVPRCWJZQK-UHFFFAOYSA-N | |
| 25612 | |
| C=CCCCCCCCCO |
| C10H20O | |
| MFCD00002992 | |
| 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol | |
| dec-9-en-1-ol |
Specifications
| 13019-22-2 | |
| 108°C | |
| MFCD00002992 | |
| 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol | |
| C=CCCCCCCCCO | |
| 156.269 | |
| 156.27 | |
| Liquid |
| Yellow | |
| C10H20O | |
| 25 mL | |
| QGFSQVPRCWJZQK-UHFFFAOYSA-N | |
| dec-9-en-1-ol | |
| 25612 | |
| ≥97.0% (GC) | |
| 9-Decen-1-ol |
Safety and Handling
EINECSNumber : (2)-0258
RTECSNumber : HE2095000
TSCA : Yes