accessibility menu, dialog, popup

UserName

HOCH2

CAS RN 13019-22-2

IUPAC Name: dec-9-en-1-ol
Synonyms: 9-decen-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: QGFSQVPRCWJZQK-UHFFFAOYSA-N
SMILES: OCCCCCCCCC=C
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)