missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol 97.0+%, TCI America™

Catalog No. D189225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D189225ML 25 mL
D1892500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D189225ML Supplier TCI America Supplier No. D189225ML

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002992
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-Decen-1-ol
EINECSNumber (2)-0258
RTECSNumber HE2095000
TSCA Yes